Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttll11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029774.2 | NP_084050.1 | 8 - 9 | 1820 | 102 | |
GenBank mRNA | AK016577.1 | - | 7 - 8 | 1178 | 102 | |
AM690755.1 | - | 8 - 9 | 1820 | 102 | ||
BC115858.1 | - | 5 - 6 | 1170 | 40057265 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |