Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cinp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026048.4 | NP_080324.1 | 2 - 3 | 246 | 65 | |
NM_027223.1 | NP_081499.1 | 2 - 3 | 246 | 65 | ||
GenBank mRNA | AK004355.1 | - | 2 - 3 | 201 | 65 | |
AK007812.1 | - | 2 - 3 | 236 | 65 | ||
AK032155.1 | - | 2 - 3 | 235 | 65 | ||
AK076126.1 | - | 2 - 3 | 216 | 65 | ||
AK160662.1 | - | 2 - 3 | 216 | 65 | ||
BC011066.1 | - | 2 - 3 | 216 | 3985202 | 65 | |
BC021867.1 | - | 2 - 3 | 198 | 3710025 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |