Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Setx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198033.2 | NP_932150.2 | 22 - 23 | 7539 | 163 | |
GenBank mRNA | AK129177.1 | - | 9 - 10 | 1116 | 163 | |
BC046382.1 | - | 12 - 13 | 1626 | 163 | ||
BC058109.1 | - | 10 - 11 | 1393 | 163 | ||
BC079604.1 | - | 11 - 12 | 1431 | 163 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |