Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Macrod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK160433.1 | - | 4 - 5 | 540 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |