Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200006817.mm.1 |
NM_029553.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200006817.mm.1 |
NM_029553.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200006817.mm.1 |
TC1200000978.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200006821.mm.1 |
NM_029553.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200006821.mm.1 |
NM_029553.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200006821.mm.1 |
NM_198311.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200006821.mm.1 |
NM_198311.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200006821.mm.1 |
TC1200000978.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200006818.mm.1 |
NM_029553.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200006818.mm.1 |
NM_029553.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200006818.mm.1 |
NM_198311.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200006818.mm.1 |
NM_198311.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200006818.mm.1 |
TC1200000978.mm.1 |
MTA TC |