Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttyh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB162929.1 | - | 2 - 3 | 309 | 70 | |
AF190699.1 | - | 2 - 3 | 381 | 70 | ||
AF190991.1 | - | 2 - 3 | 381 | 70 | ||
AK018148.1 | - | 2 - 3 | 373 | 70 | ||
AK031550.1 | - | 2 - 3 | 409 | 70 | ||
AK043998.1 | - | 2 - 3 | 429 | 70 | ||
AK048413.1 | - | 2 - 3 | 380 | 70 | ||
AK049903.1 | - | 1 - 2 | 115 | 70 | ||
AK081981.1 | - | 2 - 3 | 382 | 70 | ||
AK083126.1 | - | 1 - 2 | 506 | 70 | ||
BC046310.1 | - | 2 - 3 | 367 | 6307420 | 70 | |
BC065694.1 | - | 2 - 3 | 373 | 6813365 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |