Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rdh13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290411.1 | NP_001277340.1 | 4 - 5 | 620 | 74 | |
NM_175372.4 | NP_780581.1 | 4 - 5 | 587 | 74 | ||
GenBank mRNA | AK028434.1 | - | 4 - 5 | 580 | 74 | |
BC082583.1 | - | 4 - 5 | 539 | 30613493 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |