Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028360.2 | NP_082636.3 | 3 - 4 | 443 | 63 | |
GenBank mRNA | AK034075.1 | - | 3 - 4 | 433 | 63 | |
BC026866.1 | - | 1 - 2 | 87 | 63 | ||
BC037111.1 | - | 2 - 3 | 167 | 5370464 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |