Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011769.4 | NP_035899.4 | 6 - 7 | 546 | 78 | |
GenBank mRNA | AF111101.1 | - | 7 - 8 | 669 | 78 | |
AK086590.1 | - | 6 - 7 | 565 | 78 | ||
BC066196.1 | - | 6 - 7 | 546 | 30430385 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |