Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtpbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027000.4 | NP_081276.2 | 15 - 16 | 1628 | 73 | |
GenBank mRNA | AF325354.1 | - | 15 - 16 | 1612 | 73 | |
AF348208.1 | - | 15 - 16 | 1635 | 73 | ||
AK032309.1 | - | 3 - 4 | 323 | 73 | ||
AK163372.1 | - | 14 - 15 | 1448 | 73 | ||
AK166711.1 | - | 15 - 16 | 1635 | 73 | ||
BC005514.1 | - | 15 - 16 | 1609 | 73 | ||
BC025431.1 | - | 15 - 16 | 1633 | 5007174 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |