Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200017326.mm.1 |
NM_008780.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200017326.mm.1 |
NM_008780.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200017326.mm.1 |
TC0200002184.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200017328.mm.1 |
NM_008780.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200017328.mm.1 |
NM_008780.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200017328.mm.1 |
TC0200002184.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200017324.mm.1 |
NM_008780.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200017324.mm.1 |
NM_008780.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200017324.mm.1 |
TC0200002184.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200017325.mm.1 |
NM_008780.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200017325.mm.1 |
NM_008780.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200017325.mm.1 |
TC0200002184.mm.1 |
MTA TC |