Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK154760.1 | - | 11 - 12 | 1377 | 61 | |
AK161339.1 | - | 6 - 7 | 667 | 61 | ||
BC065140.1 | - | 11 - 12 | 1250 | 61 | ||
BC086671.1 | - | 11 - 12 | 1254 | 30635559 | 61 | |
D38557.1 | - | 11 - 12 | 1324 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |