Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200024213.mm.1 |
NM_008091.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024213.mm.1 |
NM_008091.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024213.mm.1 |
TC0200002935.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200024215.mm.1 |
NM_008091.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024215.mm.1 |
NM_008091.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024215.mm.1 |
TC0200002935.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200024216.mm.1 |
NM_008091.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024216.mm.1 |
NM_008091.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024216.mm.1 |
TC0200002935.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200024218.mm.1 |
NM_008091.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024218.mm.1 |
NM_008091.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024218.mm.1 |
TC0200002935.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200024214.mm.1 |
NM_008091.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024214.mm.1 |
NM_008091.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024214.mm.1 |
TC0200002935.mm.1 |
MTA TC |