Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163566.1 | NP_001157038.1 | 2 - 3 | 521 | 84 | |
GenBank mRNA | BC043114.1 | - | 1 - 2 | 209 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |