Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dll4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019454.3 | NP_062327.2 | 4 - 5 | 1013 | 119 | |
GenBank mRNA | AB043893.1 | - | 4 - 5 | 656 | 119 | |
AF253469.1 | - | 4 - 5 | 968 | 119 | ||
AF273454.1 | - | 4 - 5 | 1034 | 119 | ||
AK004739.1 | - | 4 - 5 | 981 | 119 | ||
AK031699.1 | - | 4 - 5 | 982 | 119 | ||
AK165517.1 | - | 4 - 5 | 982 | 119 | ||
AK171717.1 | - | 4 - 5 | 982 | 117 | ||
BC042497.1 | - | 4 - 5 | 968 | 4017786 | 119 | |
BC049130.1 | - | 4 - 5 | 973 | 6825525 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |