Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g4f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024145.2 | NP_001019316.1 | 13 - 14 | 1522 | 94 | |
GenBank mRNA | AB195278.1 | - | 13 - 14 | 1522 | 94 | |
AK076378.1 | - | 9 - 10 | 930 | 94 | ||
BC039947.1 | - | 13 - 14 | 1371 | 94 | ||
BC145426.1 | - | 13 - 14 | 1474 | 9053984 | 94 | |
BC145428.1 | - | 13 - 14 | 1474 | 9053986 | 94 | |
BC150771.1 | - | 13 - 14 | 1474 | 9087682 | 94 | |
DQ904008.1 | - | 13 - 14 | 1522 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |