Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g4f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024145.2 | NP_001019316.1 | 15 - 16 | 1776 | 55 | |
GenBank mRNA | AB195278.1 | - | 15 - 16 | 1776 | 55 | |
BC039947.1 | - | 15 - 16 | 1625 | 55 | ||
BC046400.1 | - | 2 - 3 | 219 | 55 | ||
BC145426.1 | - | 15 - 16 | 1728 | 9053984 | 55 | |
BC145427.1 | - | 15 - 16 | 1723 | 55 | ||
BC145428.1 | - | 15 - 16 | 1728 | 9053986 | 55 | |
BC150771.1 | - | 15 - 16 | 1728 | 9087682 | 55 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |