Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zscan5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133204.2 | NP_573467.2 | 2 - 3 | 230 | 131 | |
GenBank mRNA | AK139659.1 | - | 2 - 3 | 231 | 131 | |
BC139442.1 | - | 1 - 2 | 120 | 8862464 | 131 | |
BC139456.1 | - | 1 - 2 | 120 | 8862478 | 131 | |
BY725521.1 | - | 2 - 3 | 231 | 131 | ||
CN717865.1 | - | 2 - 3 | 207 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |