Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700019478.mm.1 |
TC0700002143.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700019478.mm.1 |
NM_001113474.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019478.mm.1 |
NM_001113474.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019478.mm.1 |
NM_178611.6.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019478.mm.1 |
NM_178611.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019468.mm.1 |
TC0700002143.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700019468.mm.1 |
NM_001113474.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019468.mm.1 |
NM_001113474.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019470.mm.1 |
TC0700002143.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700019470.mm.1 |
NM_001113474.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019470.mm.1 |
NM_001113474.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019470.mm.1 |
NM_178611.6.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019470.mm.1 |
NM_178611.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019471.mm.1 |
TC0700002143.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700019471.mm.1 |
NM_001113474.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019471.mm.1 |
NM_001113474.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019471.mm.1 |
NM_178611.6.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019471.mm.1 |
NM_178611.6 |
Refseq |