Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700012321.mm.1 |
NM_198958.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012321.mm.1 |
NM_198958.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012321.mm.1 |
TC1700001358.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700012322.mm.1 |
NM_198958.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012322.mm.1 |
NM_198958.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012322.mm.1 |
TC1700001358.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700012319.mm.1 |
NM_198958.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012319.mm.1 |
NM_198958.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012319.mm.1 |
TC1700001358.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700012320.mm.1 |
NM_198958.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012320.mm.1 |
NM_198958.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012320.mm.1 |
TC1700001358.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700012318.mm.1 |
NM_198958.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012318.mm.1 |
NM_198958.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012318.mm.1 |
TC1700001358.mm.1 |
MTA TC |