Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200016799.mm.1 |
NM_011625.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200016799.mm.1 |
NM_011625.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200016799.mm.1 |
TC1200002416.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200016806.mm.1 |
TC1200002416.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200016803.mm.1 |
NM_011625.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200016803.mm.1 |
NM_011625.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200016803.mm.1 |
TC1200002416.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200016802.mm.1 |
NM_011625.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200016802.mm.1 |
NM_011625.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200016802.mm.1 |
TC1200002416.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200016800.mm.1 |
NM_011625.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200016800.mm.1 |
NM_011625.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200016800.mm.1 |
TC1200002416.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200016804.mm.1 |
NM_011625.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200016804.mm.1 |
NM_011625.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200016804.mm.1 |
TC1200002416.mm.1 |
MTA TC |