Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ak7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030187.1 | NP_084463.1 | 3 - 4 | 582 | 78 | |
GenBank mRNA | AK019664.1 | - | 3 - 4 | 447 | 78 | |
AK029555.1 | - | 3 - 4 | 583 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |