Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ak7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030187.1 | NP_084463.1 | 9 - 10 | 1126 | 91 | |
GenBank mRNA | AK019664.1 | - | 9 - 10 | 991 | 91 | |
AK029555.1 | - | 9 - 10 | 1127 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |