Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190817.1 | NP_001177746.1 | 2 - 3 | 643 | 139 | |
NM_007869.3 | NP_031895.1 | 2 - 3 | 643 | 139 | ||
GenBank mRNA | AK133582.1 | - | 2 - 3 | 627 | 139 | |
AK144228.1 | - | 2 - 3 | 599 | 139 | ||
BC080300.1 | - | 2 - 3 | 624 | 139 | ||
L16953.1 | - | 2 - 3 | 320 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |