Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Caprin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111289.1 | NP_001104759.1 | 5 - 6 | 812 | 82 | |
NM_001111290.1 | NP_001104760.1 | 5 - 6 | 812 | 82 | ||
NM_001111291.1 | NP_001104761.1 | 6 - 7 | 852 | 82 | ||
NM_001111292.1 | NP_001104762.1 | 5 - 6 | 812 | 82 | ||
NM_016739.3 | NP_058019.2 | 6 - 7 | 852 | 82 | ||
GenBank mRNA | AB373955.1 | - | 5 - 6 | 674 | 82 | |
AK133891.1 | - | 6 - 7 | 814 | 82 | ||
AK141992.1 | - | 5 - 6 | 754 | 82 | ||
AK147514.1 | - | 6 - 7 | 819 | 82 | ||
AK148203.1 | - | 5 - 6 | 799 | 82 | ||
AK157707.1 | - | 6 - 7 | 836 | 82 | ||
AK166461.1 | - | 5 - 6 | 770 | 82 | ||
AK168676.1 | - | 5 - 6 | 807 | 82 | ||
AK170830.1 | - | 6 - 7 | 814 | 82 | ||
AK219211.1 | - | 2 - 3 | 287 | 82 | ||
BC052427.1 | - | 6 - 7 | 831 | 6836041 | 82 | |
U27838.1 | - | 5 - 6 | 789 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |