Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clptm1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146047.2 | NP_666159.2 | 1 - 2 | 397 | 69 | |
GenBank mRNA | AK032295.1 | - | 1 - 2 | 360 | 69 | |
AK048357.1 | - | 1 - 2 | 294 | 69 | ||
AK156203.1 | - | 1 - 2 | 288 | 69 | ||
AK165939.1 | - | 1 - 2 | 257 | 69 | ||
BC026562.1 | - | 1 - 2 | 98 | 5028264 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |