Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpcat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145376.5 | NP_663351.3 | 6 - 7 | 727 | 70 | |
GenBank mRNA | AB244717.2 | - | 6 - 7 | 727 | 70 | |
AK046079.1 | - | 4 - 5 | 522 | 70 | ||
AK046507.1 | - | 4 - 5 | 522 | 70 | ||
AK079946.1 | - | 885 | 70 | |||
AK081865.1 | - | 6 - 7 | 885 | 70 | ||
AK132379.1 | - | 6 - 7 | 861 | 70 | ||
AK155286.1 | - | 6 - 7 | 891 | 70 | ||
AK169190.1 | - | 6 - 7 | 860 | 70 | ||
AK170723.1 | - | 6 - 7 | 880 | 70 | ||
AK171582.1 | - | 6 - 7 | 880 | 70 | ||
BC005662.1 | - | 4 - 5 | 257 | 70 | ||
BC066809.1 | - | 6 - 7 | 676 | 30475544 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |