Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010020.3 | NP_034150.1 | 1 - 2 | 109 | 72 | |
GenBank mRNA | AK032629.1 | - | 1 - 2 | 67 | 72 | |
AK136885.1 | - | 1 - 2 | 72 | |||
BC054119.1 | - | 1 - 2 | 109 | 6588897 | 72 | |
U16265.1 | - | 1 - 2 | 37 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |