Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010574.2 | NP_034704.1 | 2 - 3 | 737 | 64 | |
GenBank mRNA | AF165986.1 | - | 2 - 3 | 641 | 64 | |
AF295369.1 | - | 2 - 3 | 653 | 64 | ||
AK018266.1 | - | 1 - 2 | 299 | 64 | ||
AK148520.1 | - | 2 - 3 | 896 | 64 | ||
BC029750.1 | - | 2 - 3 | 737 | 3592499 | 64 | |
BC085291.1 | - | 2 - 3 | 918 | 5347048 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |