Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Haus8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163042.1 | NP_001156514.1 | 7 - 8 | 763 | 96 | |
NM_029621.3 | NP_083897.2 | 7 - 8 | 760 | 96 | ||
GenBank mRNA | AK010398.1 | - | 7 - 8 | 548 | 96 | |
AK161850.1 | - | 522 | 96 | |||
BC003935.1 | - | 7 - 8 | 531 | 3486472 | 96 | |
BQ177845.1 | - | 2 - 3 | 177 | 96 | ||
DV651534.1 | - | 7 - 8 | 670 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |