Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25955959 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25955958 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25955960 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2035951 |
NM_029621.3 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2035951 |
AI837612.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2035951 |
AK010398.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2035951 |
AK161850.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2035951 |
NM_001163042.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
704802 |
AK040386.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
172747 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
310738 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
842496 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
791147 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
444332 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
914364 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
406600 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
255067 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
207011 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
685300 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
763535 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
692625 |
AY078982.3 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
692625 |
AY145850.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
692625 |
BC104324.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
692625 |
BC104325.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
692625 |
NM_145712.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
22081 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
102665 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
174623 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P123090 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P144693 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2035951 |
NM_029621.3 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2035951 |
AI837612.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2035951 |
AK010398.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2035951 |
AK161850.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2035951 |
NM_001163042.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25955959 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25955948 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25955953 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25955951 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25955950 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25955947 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25955957 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25955960 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25955945 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25955954 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25955961 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25955958 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25955956 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P123090 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06550473 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06550494 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P144693 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P249544 |
NM_029621.3 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P249544 |
AI837612.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P249544 |
AK010398.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P249544 |
AK161850.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P249544 |
NM_001163042.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P115265 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P123090 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P144693 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06696660 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06696662 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06696668 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25955958 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25955960 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25955959 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955932 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955947 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955873 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955921 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955958 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955871 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955902 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955956 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955897 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955926 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955877 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955896 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955899 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955953 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955912 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955904 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955927 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955948 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955885 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955848 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955846 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955857 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955907 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955922 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955889 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955891 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955879 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955906 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955888 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955914 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955957 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955929 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955851 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955844 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955887 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955924 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955915 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955911 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955934 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955841 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955960 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955855 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955875 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955905 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955874 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955951 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25955850 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25955956 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25955948 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25955953 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25955945 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25955930 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25955932 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25955950 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06753872 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07190517 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06696660 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25955947 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25955958 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25955954 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25955952 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25955937 |
- |
- |