Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013482.2 | NP_038510.2 | 10 - 11 | 1050 | 65 | |
GenBank mRNA | L08967.1 | - | 10 - 11 | 1026 | 65 | |
L10627.1 | - | 10 - 11 | 1061 | 65 | ||
L29788.1 | - | 10 - 11 | 1022 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Protein Kinase Mus musculus Probe spans exons |