Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013482.2 | NP_038510.2 | 1 - 2 | 134 | 65 | |
GenBank mRNA | BC053392.1 | - | 1 - 2 | 70 | 30075322 | 65 |
CJ044967.1 | - | 1 - 2 | 135 | 65 | ||
L08967.1 | - | 1 - 2 | 110 | 65 | ||
L10627.1 | - | 1 - 2 | 139 | 65 | ||
L29788.1 | - | 1 - 2 | 106 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |