Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013482.2 | NP_038510.2 | 5 - 6 | 551 | 94 | |
GenBank mRNA | BC053392.1 | - | 5 - 6 | 30075322 | 94 | |
L10627.1 | - | 5 - 6 | 556 | 94 | ||
L29788.1 | - | 5 - 6 | 523 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |