Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290476.1 | NP_001277405.1 | 3 - 4 | 541 | 68 | |
NM_026167.4 | NP_080443.1 | 5 - 6 | 560 | 68 | ||
GenBank mRNA | AK004677.1 | - | 5 - 6 | 532 | 68 | |
AK122491.1 | - | 3 - 4 | 541 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |