Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK139877.1 | - | 4 - 5 | 475 | 88 | |
BC049191.1 | - | 13 - 14 | 1894 | 6476607 | 88 | |
BC068151.1 | - | 12 - 13 | 2090 | 30534873 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |