Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK033628.1 | - | 3 - 4 | 350 | 132 | |
AK139877.1 | - | 8 - 9 | 1000 | 132 | ||
BC049191.1 | - | 17 - 18 | 2419 | 6476607 | 132 | |
BC068151.1 | - | 16 - 17 | 2615 | 30534873 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |