Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290783.1 | NP_001277712.1 | 7 - 8 | 1653 | 99 | |
NM_026137.5 | NP_080413.2 | 7 - 8 | 1687 | 99 | ||
NM_028542.1 | NP_082818.1 | 7 - 8 | 1939 | 99 | ||
GenBank mRNA | AF220146.1 | - | 6 - 7 | 1209 | 99 | |
AF353243.1 | - | 7 - 8 | 1333 | 99 | ||
AF513713.1 | - | 7 - 8 | 1156 | 99 | ||
AK006837.1 | - | 6 - 7 | 1175 | 99 | ||
AK017549.1 | - | 7 - 8 | 1269 | 99 | ||
BC057366.1 | - | 7 - 8 | 1653 | 99 | ||
BC080853.1 | - | 7 - 8 | 1602 | 99 | ||
BC108332.1 | - | 7 - 8 | 1443 | 6493226 | 99 | |
BC120553.1 | - | 7 - 8 | 1190 | 40129476 | 99 | |
BC125287.1 | - | 7 - 8 | 1189 | 40129802 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |