Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfe3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105196.1 | NP_001098666.1 | 2 - 3 | 514 | 61 | |
NM_001105197.1 | NP_001098667.1 | 2 - 3 | 200 | 61 | ||
NM_001271490.1 | NP_001258419.1 | 2 - 3 | 667 | 61 | ||
NM_001271491.1 | NP_001258420.1 | 2 - 3 | 234 | 61 | ||
NM_172472.3 | NP_766060.2 | 2 - 3 | 514 | 61 | ||
GenBank mRNA | AF510653.1 | - | 2 - 3 | 356 | 61 | |
AK139756.1 | - | 2 - 3 | 511 | 61 | ||
AK144657.1 | - | 2 - 3 | 479 | 61 | ||
AK145029.1 | - | 2 - 3 | 633 | 61 | ||
AK147405.1 | - | 1 - 2 | 969 | 61 | ||
AK150372.1 | - | 2 - 3 | 267 | 61 | ||
AK151447.1 | - | 2 - 3 | 185 | 61 | ||
AK152221.1 | - | 2 - 3 | 185 | 61 | ||
AK153051.1 | - | 2 - 3 | 267 | 61 | ||
AK154690.1 | - | 2 - 3 | 480 | 61 | ||
AK155883.1 | - | 2 - 3 | 236 | 61 | ||
BC032963.1 | - | 2 - 3 | 395 | 61 | ||
BY208269.1 | - | 2 - 3 | 170 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |