Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P32240056 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P32240054 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P32240057 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P32240059 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P32240058 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P32240055 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2099379 |
BC063047.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2099379 |
S76673.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2099379 |
BC056358.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2099379 |
AK155883.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2099379 |
AK159162.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2099378 |
BC063047.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2099378 |
BC056358.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2099378 |
AK159162.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2099378 |
AK152221.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2099378 |
AK154690.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
108057 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
268443 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
327182 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
493129 |
AK052601.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
493129 |
AK088355.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
493129 |
AK090024.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
493129 |
AK146006.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
493129 |
AK172310.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
493129 |
AK206905.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
493129 |
AK207905.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
493129 |
BC024616.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
493129 |
NM_153570.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
173060 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
911287 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
58055 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
4956 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
113892 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P135188 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P141616 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2099379 |
BC063047.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2099379 |
S76673.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2099379 |
BC056358.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2099379 |
AK155883.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2099379 |
AK159162.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2099378 |
BC063047.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2099378 |
BC056358.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2099378 |
AK159162.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2099378 |
AK152221.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2099378 |
AK154690.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P461313 |
BC063047.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P461313 |
BC051568.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P461313 |
BC056358.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P461313 |
BC032963.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P461313 |
AK155883.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P199671 |
BC063047.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P199671 |
S76673.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P199671 |
BC056358.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P199671 |
AK159162.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P199671 |
AK152221.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P34397 |
S57655.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32240077 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32240049 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32240038 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32240068 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32240059 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32240076 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32240053 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32240056 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32240072 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32240070 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32240058 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32240045 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32240051 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32240066 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32240054 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32240061 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32240039 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32240067 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32240052 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32240069 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32240057 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32240074 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32240042 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32240047 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32240062 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32240040 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32240036 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P135188 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P03635391 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P141616 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P03635377 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P34397 |
S57655.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P199671 |
BC063047.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P199671 |
S76673.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P199671 |
BC056358.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P199671 |
AK159162.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P199671 |
AK152221.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P461313 |
BC063047.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P461313 |
BC051568.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P461313 |
BC056358.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P461313 |
BC032963.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P461313 |
AK155883.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P131170 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P141616 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P135188 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P03635377 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14650620 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P15175273 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14650619 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P15175279 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14650616 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P32240054 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P32240057 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P32240056 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P32240058 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P32240059 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P32240055 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240058 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240028 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240037 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240119 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240108 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240051 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240126 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240036 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240083 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240109 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240138 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240072 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240161 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240080 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240090 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240121 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240117 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240162 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240116 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240034 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240141 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240142 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240129 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240127 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240152 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240059 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240114 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240097 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240099 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240033 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240039 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240092 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240125 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240076 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240074 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240078 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240111 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240100 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240131 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240035 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240066 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240135 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240133 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240145 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240047 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240095 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240158 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240106 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240103 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240068 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240148 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240070 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240157 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240140 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240105 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240136 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240052 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240079 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240038 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240124 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240054 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240056 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240031 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240040 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240049 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240045 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240069 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240089 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240093 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240134 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240085 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240087 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240062 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240123 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240046 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240041 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240156 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240101 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240086 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240042 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32240137 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240117 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240036 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240131 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240129 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240134 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240062 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240093 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240123 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240074 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240135 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P15175279 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P15051882 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14897468 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240101 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P15058457 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240033 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240034 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240092 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240041 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240031 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240067 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240114 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240030 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P15058475 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P15175263 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240130 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240112 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P15017483 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240049 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240090 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240109 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240052 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240068 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240046 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240080 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240037 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240099 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14855551 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P15175269 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240106 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240056 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240053 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240051 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240066 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240125 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240043 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240078 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240089 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14973299 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P15060375 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240070 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P15058462 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240076 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240111 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240126 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240121 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240087 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240103 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240042 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14650616 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240083 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P15175275 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240040 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P15017477 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240038 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240079 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240127 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14693942 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240124 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14667851 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32240039 |
- |
- |