Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Palld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081390.1 | NP_001074859.1 | 17 - 18 | 3249 | 85 | |
NM_001293772.1 | NP_001280701.1 | 19 - 20 | 3979 | 85 | ||
NM_001293773.1 | NP_001280702.1 | 17 - 18 | 3059 | 85 | ||
NM_001293774.1 | NP_001280703.1 | 9 - 10 | 1825 | 85 | ||
GenBank mRNA | AF205078.1 | - | 7 - 8 | 1003 | 85 | |
AK010350.1 | - | 5 - 6 | 712 | 85 | ||
AK031696.1 | - | 17 - 18 | 3052 | 85 | ||
AK052489.1 | - | 19 - 20 | 3962 | 85 | ||
AK076423.1 | - | 16 - 17 | 2697 | 85 | ||
AK087573.1 | - | 9 - 10 | 1201 | 85 | ||
AK149663.1 | - | 9 - 10 | 1802 | 85 | ||
AK173081.1 | - | 17 - 18 | 2999 | 85 | ||
BC027364.1 | - | 17 - 18 | 3008 | 85 | ||
BC076588.1 | - | 10 - 11 | 2087 | 85 | ||
BC127081.1 | - | 10 - 11 | 1833 | 85 | ||
JX477684.1 | - | 17 - 18 | 3051 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |