Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anks6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC060663.1 | - | 8 - 9 | 1617 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |