Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oog4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173773.1 | NP_776134.1 | 1 - 2 | 47 | 77 | |
GenBank mRNA | AK054346.1 | - | 1 - 2 | 69 | 77 | |
AY573564.1 | - | 1 - 2 | 47 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |