Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008823.3 | NP_032849.2 | 4 - 5 | 629 | 92 | |
GenBank mRNA | AK150212.1 | - | 4 - 5 | 590 | 92 | |
AK152782.1 | - | 4 - 5 | 580 | 92 | ||
AK154306.1 | - | 4 - 5 | 613 | 92 | ||
AK159291.1 | - | 4 - 5 | 630 | 92 | ||
AK159609.1 | - | 4 - 5 | 92 | |||
AK159853.1 | - | 4 - 5 | 92 | |||
AK171374.1 | - | 4 - 5 | 613 | 92 | ||
BC132369.1 | - | 4 - 5 | 613 | 40130646 | 92 | |
BC138306.1 | - | 4 - 5 | 613 | 8861326 | 92 | |
X12905.1 | - | 3 - 4 | 478 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |