Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stt3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024222.2 | NP_077184.2 | 10 - 11 | 1901 | 75 | |
GenBank mRNA | AK012153.1 | - | 10 - 11 | 1571 | 75 | |
AK018758.2 | - | 10 - 11 | 1611 | 75 | ||
AK145674.1 | - | 10 - 11 | 1542 | 75 | ||
AK148065.1 | - | 10 - 11 | 1599 | 75 | ||
AK152899.1 | - | 10 - 11 | 1949 | 75 | ||
AK154979.1 | - | 10 - 11 | 1590 | 75 | ||
AK170079.1 | - | 10 - 11 | 1613 | 75 | ||
AX799084.1 | - | 10 - 11 | 1612 | 75 | ||
BC003206.1 | - | 5 - 6 | 613 | 75 | ||
BC052433.1 | - | 10 - 11 | 1901 | 6837097 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |