Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gadl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028638.1 | NP_082914.1 | 3 - 4 | 362 | 82 | |
GenBank mRNA | BC052327.1 | - | 3 - 4 | 362 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |