Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175114.3 | NP_780323.2 | 3 - 4 | 505 | 56 | |
GenBank mRNA | AK029123.1 | - | 4 - 5 | 903 | 56 | |
AK041436.1 | - | 4 - 5 | 873 | 56 | ||
AK052264.1 | - | 4 - 5 | 892 | 56 | ||
AK122426.1 | - | 4 - 5 | 595 | 56 | ||
BC046285.1 | - | 3 - 4 | 495 | 56 | ||
BC058971.1 | - | 3 - 4 | 447 | 6832507 | 56 | |
BC116262.1 | - | 3 - 4 | 472 | 56 | ||
BC116263.1 | - | 3 - 4 | 472 | 56 | ||
BC129961.1 | - | 3 - 4 | 472 | 56 | ||
BC129962.1 | - | 3 - 4 | 472 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |