Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198664.3 | NP_941066.3 | 9 - 10 | 2299 | 86 | |
GenBank mRNA | AK148254.1 | - | 9 - 10 | 2302 | 86 | |
AK148289.1 | - | 9 - 10 | 2215 | 86 | ||
BE656277.1 | - | 1 - 2 | 94 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |