Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198664.3 | NP_941066.3 | 11 - 12 | 2602 | 62 | |
GenBank mRNA | AK148289.1 | - | 11 - 12 | 2518 | 62 | |
BE656277.1 | - | 3 - 4 | 397 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |