Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coro2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164804.1 | NP_001158276.1 | 7 - 8 | 986 | 62 | |
NM_178893.4 | NP_849224.2 | 8 - 9 | 1211 | 62 | ||
GenBank mRNA | AK030097.1 | - | 8 - 9 | 1214 | 62 | |
AK030971.1 | - | 7 - 8 | 943 | 62 | ||
AK044060.1 | - | 7 - 8 | 948 | 62 | ||
AK054394.1 | - | 7 - 8 | 987 | 62 | ||
AK152490.1 | - | 7 - 8 | 975 | 62 | ||
AK153134.1 | - | 7 - 8 | 975 | 62 | ||
AK154247.1 | - | 7 - 8 | 943 | 62 | ||
AK170184.1 | - | 7 - 8 | 944 | 62 | ||
BC031164.1 | - | 5 - 6 | 633 | 62 | ||
BC138731.1 | - | 7 - 8 | 1011 | 8861753 | 62 | |
BC138732.1 | - | 7 - 8 | 1011 | 8861754 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |